Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 4-acetylbenzoate, 98%

Chemical Identifiers

Chemical Name or Material Ethyl 4-acetylbenzoate
CAS 38430-55-6
Formula Weight 192.21
MDL Number MFCD00013241
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6325803

  SDS
alfa aesar™
H6325803
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6325806

  SDS
alfa aesar™
H6325806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 192.21
Melting Point 55° to 57°C
CAS 38430-55-6
Quantity 1g
Physical Form Solid
Assay 98%
MDL Number MFCD00013241
Chemical Name or Material Ethyl 4-acetylbenzoate