Alfa Aesar™ Ethyl 4-acetylbenzoate, 98%
Manufacturer:  Alfa Aesar™ H6325806
 View more versions of this product
Catalog No. AAH6325806
Specifications
Specifications
Ethyl 4-acetylbenzoate
5g
C11H12O3
4-acetylbenzoic acid ethyl ester, benzoic acid, 4-acetyl-, ethyl ester, 4-acetyl-benzoic acid ethyl ester, ethyl4-acetylbenzoate, ethyl 4-acetylbenzate, pubchem18162, ethyl 4-acetyl benzoate, ethyl 4-acetyl-benzoate, acmc-1csm7, ethyl 4-acetylbenzoate
CCOC(=O)C1=CC=C(C=C1)C(=O)C
192.214
192.21
98%
55°C to 57°C
38430-55-6
MFCD00013241
GLOAPLPTWAXAIG-UHFFFAOYSA-N
ethyl 4-acetylbenzoate
600911
Solid
Chemical Identifiers
38430-55-6
192.214
GLOAPLPTWAXAIG-UHFFFAOYSA-N
600911
CCOC(=O)C1=CC=C(C=C1)C(=O)C
C11H12O3
MFCD00013241
4-acetylbenzoic acid ethyl ester, benzoic acid, 4-acetyl-, ethyl ester, 4-acetyl-benzoic acid ethyl ester, ethyl4-acetylbenzoate, ethyl 4-acetylbenzate, pubchem18162, ethyl 4-acetyl benzoate, ethyl 4-acetyl-benzoate, acmc-1csm7, ethyl 4-acetylbenzoate
ethyl 4-acetylbenzoate
SDS Documents