Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 4-(5-formyl-2-furyl)benzoate, 97%

$296.12 - $296.12

Chemical Identifiers

Chemical Name or Material Ethyl 4-(5-formyl-2-furyl)benzoate
CAS 19247-87-1
Molecular Formula C14H12O4
Formula Weight 244.25g/mol
MDL Number MFCD08461682
Synonym 5-(4-Ethoxycarbonylphenyl)-2-furaldehyde; 4-(5-Formyl-2-furyl)benzoic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5048406

 
alfa aesar™
H50484-06
Each for $296.12
Description & Specifications

Specifications

Quantity 5g
Chemical Name or Material Ethyl 4-(5-formyl-2-furyl)benzoate
CAS 19247-87-1
Assay 97%
Molecular Formula C14H12O4
Formula Weight 244.25g/mol
MDL Number MFCD08461682
Synonym 5-(4-Ethoxycarbonylphenyl)-2-furaldehyde; 4-(5-Formyl-2-furyl)benzoic acid ethyl ester