Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 4,5-dimethoxy-2-nitrobenzoate, 98+%

$87.28 - $87.28

Chemical Identifiers

Chemical Name or Material Ethyl 4,5-dimethoxy-2-nitrobenzoate
CAS 100905-33-7
Molecular Formula C11H13NO6
Formula Weight 255.23
MDL Number MFCD00015474
Beilstein 2699210
Synonym 4,5-Dimethoxy-2-nitrobenzoic acid ethyl ester; Ethyl 3,4-dimethoxy-6-nitrobenzoate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0252409

 
alfa aesar™
L0252409
10g Each for $87.28
Description & Specifications

Specifications

Formula Weight 255.23
Chemical Name or Material Ethyl 4,5-dimethoxy-2-nitrobenzoate
Assay ≥98%
MDL Number MFCD00015474
Molecular Formula C11H13NO6
CAS 100905-33-7
Beilstein 2699210
Synonym 4,5-Dimethoxy-2-nitrobenzoic acid ethyl ester; Ethyl 3,4-dimethoxy-6-nitrobenzoate
Melting Point 100° to 102°C
Quantity 10g