missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl 4,4-Difluoroacetoacetate, TCI America™

Manufacturer:  TCI America E10185G

 View more versions of this product

Catalog No. E10185G



Specifications

Specifications

Ethyl 4,4-Difluoroacetoacetate
Yellow
5g
C6H8F2O3
ethyl 4,4-difluoroacetoacetate, ethyl 4,4-difluoro-3-oxobutyrate, ethyl difluoroacetoacetate, butanoic acid, 4,4-difluoro-3-oxo-, ethyl ester, ethyl4,4-difluoro-3-oxobutyrate, ethyl 4,4-difluoro-3-oxo-butanoate, 4,4-difluoroacetoacetic acid ethyl ester, 4,4-difluoro-3-oxo-butyric acid ethyl ester, ethyl4,4-difluoro-3-oxobutanoate, pubchem21933
CCOC(=O)CC(=O)C(F)F
166.124
166.12
≥96.0% (GC)
160°C
-46°C
352-24-9
MFCD00039277
CBDPWKVOPADMJC-UHFFFAOYSA-N
ethyl 4,4-difluoro-3-oxobutanoate
67697
Clear Liquid at 20°C

Chemical Identifiers

352-24-9
166.124
CBDPWKVOPADMJC-UHFFFAOYSA-N
67697
CCOC(=O)CC(=O)C(F)F
C6H8F2O3
MFCD00039277
ethyl 4,4-difluoroacetoacetate, ethyl 4,4-difluoro-3-oxobutyrate, ethyl difluoroacetoacetate, butanoic acid, 4,4-difluoro-3-oxo-, ethyl ester, ethyl4,4-difluoro-3-oxobutyrate, ethyl 4,4-difluoro-3-oxo-butanoate, 4,4-difluoroacetoacetic acid ethyl ester, 4,4-difluoro-3-oxo-butyric acid ethyl ester, ethyl4,4-difluoro-3-oxobutanoate, pubchem21933
ethyl 4,4-difluoro-3-oxobutanoate
Safety and Handling

Safety and Handling

TSCA : Yes

Recommended Storage : Refrigerator

Documents