Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 4,4,4-trifluoro-3-hydroxybutyrate, 97%

$121.30 - $466.41

Chemical Identifiers

Chemical Name or Material Ethyl 4,4,4-trifluoro-3-hydroxybutyrate
CAS 372-30-5
Molecular Formula C6H9F3O3
Formula Weight 186.13
MDL Number MFCD00040842
Beilstein 1708105
Synonym 4,4,4-Trifluoro-3-hydroxybutyric acid ethyl ester; Ethyl 3-hydroxy-4,4,4-trifluorobutyrate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1275309

 
alfa aesar™
L1275309
10g Each for $121.30

AAL1275318

 
alfa aesar™
L1275318
50g Each for $466.41
Description & Specifications

Specifications

Formula Weight 186.13
Chemical Name or Material Ethyl 4,4,4-trifluoro-3-hydroxybutyrate
Molecular Formula C6H9F3O3
Assay 97%
MDL Number MFCD00040842
Beilstein 1708105
Synonym 4,4,4-Trifluoro-3-hydroxybutyric acid ethyl ester; Ethyl 3-hydroxy-4,4,4-trifluorobutyrate
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Boiling Point 90° to 91°C (50mmHg)
Melting Point 23°C
Quantity 10g
CAS 372-30-5
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.