missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 4,4,4-trifluoro-3-hydroxybutyrate, 97%

Manufacturer:  Alfa Aesar™ L1275318

 View more versions of this product

Catalog No. AAL1275318



Specifications

Specifications

Ethyl 4,4,4-trifluoro-3-hydroxybutyrate
23°C
372-30-5
MFCD00040842
ethyl 3-hydroxy-4,4,4-trifluorobutyrate, ethyl 4,4,4-trifluoro-3-hydroxybutyrate, ethyl 3-hydroxy-4,4,4-trifluorobutanoate, 3-hydroxy-4,4,4-trifluorobutyric acid ethyl ester, 4,4,4-trifluoro-3-hydroxybutyric acid ethyl ester, ethyl3-hydroxy-4,4,4-trifluorobutyrate, butanoic acid, 4,4,4-trifluoro-3-hydroxy-, ethyl ester, acmc-209irs, acmc-20al28, ksc496k8b
CCOC(=O)CC(C(F)(F)F)O
186.13
186.13
90°C to 91°C (50mmHg)
50g
C6H9F3O3
1708105
ZWEDFBKLJILTMC-UHFFFAOYSA-N
ethyl 4,4,4-trifluoro-3-hydroxybutanoate
136210
97%

Chemical Identifiers

372-30-5
186.13
ZWEDFBKLJILTMC-UHFFFAOYSA-N
136210
CCOC(=O)CC(C(F)(F)F)O
C6H9F3O3
MFCD00040842
ethyl 3-hydroxy-4,4,4-trifluorobutyrate, ethyl 4,4,4-trifluoro-3-hydroxybutyrate, ethyl 3-hydroxy-4,4,4-trifluorobutanoate, 3-hydroxy-4,4,4-trifluorobutyric acid ethyl ester, 4,4,4-trifluoro-3-hydroxybutyric acid ethyl ester, ethyl3-hydroxy-4,4,4-trifluorobutyrate, butanoic acid, 4,4,4-trifluoro-3-hydroxy-, ethyl ester, acmc-209irs, acmc-20al28, ksc496k8b
ethyl 4,4,4-trifluoro-3-hydroxybutanoate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning

Documents