missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 4,4,4-trifluoro-2-methylacetoacetate, 96%

$41.64 - $115.35

Chemical Identifiers

CAS 344-00-3
Molecular Formula C7H9F3O3
Molecular Weight (g/mol) 198.141
MDL Number MFCD00190635
InChI Key YLRGPBKEZVHOAW-UHFFFAOYSA-N
Synonym ethyl 2-methyl-4,4,4-trifluoroacetoacetate, butanoic acid, 4,4,4-trifluoro-2-methyl-3-oxo-, ethyl ester, zlchem 125, acmc-20al0w, ethyl 2-methyltrifluoroacetoacetate, ethyl 2-methyl-4,4,4-trifluoro acetoacetate, ethyl alpha-methyl-4,4,4-trifluoroacetoacetate, ethyl4,4,4-trifluoro-2-methyl-3-oxobutanoate, ethyl 3-keto-2-methyl-4,4,4-trifluorobutanoate, ethyl 4,4,4-trifluoro-2-methyl-3-oxo-butanoate
PubChem CID 2737204
IUPAC Name ethyl 4,4,4-trifluoro-2-methyl-3-oxobutanoate
SMILES CCOC(=O)C(C)C(=O)C(F)(F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1733503 View Documents Alfa Aesar™
L1733503
1g Each for $41.64
 
AAL1733506 View Documents Alfa Aesar™
L1733506
5g Each for $115.35
 
Specifications

Chemical Identifiers

344-00-3
198.141
YLRGPBKEZVHOAW-UHFFFAOYSA-N
2737204
CCOC(=O)C(C)C(=O)C(F)(F)F
C7H9F3O3
MFCD00190635
ethyl 2-methyl-4,4,4-trifluoroacetoacetate, butanoic acid, 4,4,4-trifluoro-2-methyl-3-oxo-, ethyl ester, zlchem 125, acmc-20al0w, ethyl 2-methyltrifluoroacetoacetate, ethyl 2-methyl-4,4,4-trifluoro acetoacetate, ethyl alpha-methyl-4,4,4-trifluoroacetoacetate, ethyl4,4,4-trifluoro-2-methyl-3-oxobutanoate, ethyl 3-keto-2-methyl-4,4,4-trifluorobutanoate, ethyl 4,4,4-trifluoro-2-methyl-3-oxo-butanoate
ethyl 4,4,4-trifluoro-2-methyl-3-oxobutanoate

Specifications

47°C (116°F)
1g
C7H9F3O3
UN3272
ethyl 2-methyl-4,4,4-trifluoroacetoacetate, butanoic acid, 4,4,4-trifluoro-2-methyl-3-oxo-, ethyl ester, zlchem 125, acmc-20al0w, ethyl 2-methyltrifluoroacetoacetate, ethyl 2-methyl-4,4,4-trifluoro acetoacetate, ethyl alpha-methyl-4,4,4-trifluoroacetoacetate, ethyl4,4,4-trifluoro-2-methyl-3-oxobutanoate, ethyl 3-keto-2-methyl-4,4,4-trifluorobutanoate, ethyl 4,4,4-trifluoro-2-methyl-3-oxo-butanoate
CCOC(=O)C(C)C(=O)C(F)(F)F
198.141
198.14
Ethyl 4,4,4-trifluoro-2-methylacetoacetate
144°C to 146°C
344-00-3
MFCD00190635
1783620
YLRGPBKEZVHOAW-UHFFFAOYSA-N
ethyl 4,4,4-trifluoro-2-methyl-3-oxobutanoate
2737204
96%
Safety and Handling

Safety and Handling

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.