missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate, 97+%, ACROS Organics™

Chemical Identifiers

CAS 195062-62-5
Molecular Formula C15H21BO4
Molecular Weight (g/mol) 276.14
MDL Number MFCD03453659
InChI Key NCVIYKCFTYSAGN-UHFFFAOYSA-N
Synonym 2-4-ethoxycarbonyl phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
PubChem CID 2734618
IUPAC Name ethyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)C(=O)OCC
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC366360010 SDS View Documents Thermo Fisher Scientific
AC366360010
1g Glass bottle N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

195062-62-5
276.14
NCVIYKCFTYSAGN-UHFFFAOYSA-N
2734618
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)C(=O)OCC
C15H21BO4
MFCD03453659
2-4-ethoxycarbonyl phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
ethyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate

Specifications

1.0520g/mL
Authentic
1.4980 to 1.5000 (20°C, 589nm)
330°C
1g
97% min. (GC)
MFCD03453659
Solubility in water: insoluble in water
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)C(=O)OCC
276.14
276.14
97+%
>110°C
Glass bottle
1.052
Yellow
195062-62-5
C15H21BO4
2-4-ethoxycarbonyl phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
NCVIYKCFTYSAGN-UHFFFAOYSA-N
ethyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate
2734618
Viscous Liquid
Ethyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate
Safety and Handling

Safety and Handling

Recommended Storage : Refrigerator +4°C