missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate, 97+%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC366360010

 View more versions of this product

Catalog No. AC366360010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

Ethyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate
>110°C
Glass bottle
1.052
Yellow
195062-62-5
C15H21BO4
2-4-ethoxycarbonyl phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoic acid ethyl ester, 4-carboethoxyphenylboronic acid pinacol ester, 4-ethoxycarbonyl benzeneboronic acid pinacol ester, 4-ethoxycarbonylphenylboronic acid pinacol ester, 4-ethoxycarbonylphenylboronic acid, pinacol ester, ethyl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoate, ethyl 4-tetramethyl-1,3,2-dioxaborolan-2-yl benzoate, ethyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoate
NCVIYKCFTYSAGN-UHFFFAOYSA-N
ethyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate
2734618
Viscous Liquid
1.0520g/mL
Authentic
1.4980 to 1.5000 (20°C, 589nm)
330°C
1g
97% min. (GC)
MFCD03453659
Solubility in water: insoluble in water
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)C(=O)OCC
276.14
276.14
97+%

Chemical Identifiers

195062-62-5
276.14
NCVIYKCFTYSAGN-UHFFFAOYSA-N
2734618
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)C(=O)OCC
C15H21BO4
MFCD03453659
2-4-ethoxycarbonyl phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoic acid ethyl ester, 4-carboethoxyphenylboronic acid pinacol ester, 4-ethoxycarbonyl benzeneboronic acid pinacol ester, 4-ethoxycarbonylphenylboronic acid pinacol ester, 4-ethoxycarbonylphenylboronic acid, pinacol ester, ethyl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoate, ethyl 4-tetramethyl-1,3,2-dioxaborolan-2-yl benzoate, ethyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoate
ethyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate
Safety and Handling

Safety and Handling

Recommended Storage : Refrigerator +4°C

SDS Documents