missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl 4-(2-Hydroxy-2-propyl)-2-propyl-1H-imidazole-5-carboxylate, TCI America™

Manufacturer:  TCI America E09765G

 View more versions of this product

Catalog No. E09765G



Specifications

Specifications

Ethyl 4-(2-Hydroxy-2-propyl)-2-propyl-1H-imidazole-5-carboxylate
104°C
144689-93-0
MFCD07787565
KZBJJAFGNMRRHN-UHFFFAOYSA-N
ethyl 5-(2-hydroxypropan-2-yl)-2-propyl-1H-imidazole-4-carboxylate
10562026
Crystal-Powder at 20°C
Yellow
5g
C12H20N2O3
4-(2-Hydroxy-2-propyl)-2-propyl-1H-imidazole-5-carboxylic Acid Ethyl Ester
CCCC1=NC(=C(N1)C(C)(C)O)C(=O)OCC
240.303
240.30
≥96.0% (GC,T)

Chemical Identifiers

144689-93-0
240.303
KZBJJAFGNMRRHN-UHFFFAOYSA-N
10562026
CCCC1=NC(=C(N1)C(C)(C)O)C(=O)OCC
C12H20N2O3
MFCD07787565
4-(2-Hydroxy-2-propyl)-2-propyl-1H-imidazole-5-carboxylic Acid Ethyl Ester
ethyl 5-(2-hydroxypropan-2-yl)-2-propyl-1H-imidazole-4-carboxylate
Safety and Handling

Safety and Handling

TSCA : No

Documents