Please login to your online account to display your discounted pricing

Ethyl 3-(trimethylsilyl)propiolate, 99%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC381550250

 View more versions of this product

Catalog No. AC381550250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Assay ≥98.5%
Formula Weight 170.28
Quantity 25g
Color Clear colorless
Density 0.8970g/mL
Chemical Name or Material Ethyl 3-(trimethylsilyl)propiolate
MDL Number MFCD00074886
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Infrared Spectrum Authentic
Refractive Index 1.4410 to 1.4430 (20°C, 589nm)
Assay Percent Range 99%
Molecular Formula C8H14O2Si
Linear Formula (CH3)3SiCidentical toCCOOCH2CH3
CAS 16205-84-8
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle

Specifications

Chemical Name or Material Ethyl 3-(trimethylsilyl)propiolate
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C8H14O2Si
Linear Formula (CH3)3SiCidentical toCCOOCH2CH3
Formula Weight 170.28
MDL Number MFCD00074886
Accessories

GHS Signal Word: Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents