missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 3-(trimethylsilyl)propiolate, 98%

Manufacturer:  Alfa Aesar™ H5351714

 View more versions of this product

Catalog No. AAH5351714



Description

Description

Applications
Ethyl 3-(trimethylsilyl)propiolate was used in the preparation of 4-ethoxycarbonyl-3-(trimethylsilyl)furan.

Solubility
Sparingly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Specifications

Ethyl 3-(trimethylsilyl)propiolate
C8H14O2Si
MFCD00074886
2-propynoic acid, 3-trimethylsilyl-, ethyl ester
QRBKPHYAIRLCLK-UHFFFAOYSA-N
ethyl 3-trimethylsilylprop-2-ynoate
568601
98%
1.441
25g
16205-84-8
(CH3)3SiC≡CCO2CH2CH3
1767850
Sparingly soluble in water.
CCOC(=O)C#C[Si](C)(C)C
170.283
170.28
0.897
92°C to 96°C (28mmHg)
71°C (160°F)

Chemical Identifiers

16205-84-8
170.283
QRBKPHYAIRLCLK-UHFFFAOYSA-N
568601
CCOC(=O)C#C[Si](C)(C)C
C8H14O2Si
MFCD00074886
2-propynoic acid, 3-trimethylsilyl-, ethyl ester
ethyl 3-trimethylsilylprop-2-ynoate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

P210-P235-P261-P264b-P271-P280-P304+P340-P305+P351+P338-P312-P370+P378q-P501c

H227-H319-H335

DOTInformation : Hazard Class: 9; Packaging Group: III

TSCA : No

Recommended Storage : Ambient temperatures

Documents