Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 3-trifluoromethyl-1H-pyrazole-5-carboxylate, 97%

Chemical Identifiers

Chemical Name or Material Ethyl 3-trifluoromethyl-1H-pyrazole-5-carboxylate
CAS 129768-30-5
Formula Weight 208.14
MDL Number MFCD04111238
Synonym 3-Trifluoromethyl-1H-pyrazole-5-carboxylic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3359903

  SDS
alfa aesar™
H3359903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH33599MD

  SDS
alfa aesar™
H33599MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 208.14
Melting Point 93° to 98°C
Chemical Name or Material Ethyl 3-trifluoromethyl-1H-pyrazole-5-carboxylate
Physical Form Solid
Assay 97%
MDL Number MFCD04111238
CAS 129768-30-5
Synonym 3-Trifluoromethyl-1H-pyrazole-5-carboxylic acid ethyl ester
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Color White
Quantity 1g

Applications
Employed as a intermediate for pharmaceutical.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed containers. With adequate ventilation. Store away from oxidizing agent.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.