Learn More
Ethyl 3-pyridylacetate, 99%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 118920050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Ethyl 3-pyridylacetate | |
39931-77-6 | |
80°C (0.4mmHg) | |
Authentic | |
Glass bottle | |
1.4990 to 1.501 | |
MFCD00006411 | |
1.08 | |
RPWXYCRIAGBAGY-UHFFFAOYSA-N | |
ethyl 2-pyridin-3-ylacetate | |
96490 | |
99% |
99% | |
1.0800g/mL | |
>110°C | |
99% | |
C9H11NO2 | |
(C5H4N)CH2COOC2H5 | |
5g | |
ethyl 3-pyridylacetate, ethyl 2-pyridin-3-yl acetate, ethyl 3-pyridineacetate, 3-pyridineacetic acid, ethyl ester, acetic acid, 2-3-pyridyl-, ethyl ester, pyridin-3-yl-acetic acid ethyl ester, 3-pyridylacetic acid ethyl ester, ethyl 2-3-pyridyl acetate, pubchem20506, ethyl-3-pyridyl-acetate | |
CCOC(=O)CC1=CN=CC=C1 | |
165.19 | |
165.19 |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee
GHS Signal Word: Warning
EINECSNumber : 254-707-6
RTECSNumber : UR8950000
RUO â Research Use Only