missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 3-pyridineacetate, 99%

Manufacturer:  Alfa Aesar™ A1476806

 View more versions of this product

Catalog No. AAA1476806



Specifications

Specifications

Ethyl 3-pyridineacetate
79°C to 80°C (0.4mmHg)
39931-77-6
MFCD00006411
ethyl 3-pyridylacetate, ethyl 2-pyridin-3-yl acetate, ethyl 3-pyridineacetate, 3-pyridineacetic acid, ethyl ester, acetic acid, 2-3-pyridyl-, ethyl ester, pyridin-3-yl-acetic acid ethyl ester, 3-pyridylacetic acid ethyl ester, ethyl 2-3-pyridyl acetate, pubchem20506, ethyl-3-pyridyl-acetate
CCOC(=O)CC1=CN=CC=C1
165.192
165.19
>110°C (230°F)
5g
C9H11NO2
123827
RPWXYCRIAGBAGY-UHFFFAOYSA-N
ethyl 2-pyridin-3-ylacetate
96490
99%

Chemical Identifiers

39931-77-6
165.192
RPWXYCRIAGBAGY-UHFFFAOYSA-N
96490
CCOC(=O)CC1=CN=CC=C1
C9H11NO2
MFCD00006411
ethyl 3-pyridylacetate, ethyl 2-pyridin-3-yl acetate, ethyl 3-pyridineacetate, 3-pyridineacetic acid, ethyl ester, acetic acid, 2-3-pyridyl-, ethyl ester, pyridin-3-yl-acetic acid ethyl ester, 3-pyridylacetic acid ethyl ester, ethyl 2-3-pyridyl acetate, pubchem20506, ethyl-3-pyridyl-acetate
ethyl 2-pyridin-3-ylacetate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : UR8950000

SDS Documents