missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl 3-Piperidinecarboxylate, TCI America™

Manufacturer:  TCI America E058425G

 View more versions of this product

Catalog No. E058425G



Specifications

Specifications

Ethyl 3-Piperidinecarboxylate
Yellow
5006-62-2
MFCD00005991
XIWBSOUNZWSFKU-UHFFFAOYSA-N
ethyl piperidine-3-carboxylate
98969
Clear Liquid at 20°C
104°C
25g
C8H15NO2
ethyl nipecotate, ethyl 3-piperidinecarboxylate, nipecotic acid ethyl ester, 3-piperidinecarboxylic acid ethyl ester, ethylpiperidine-3-carboxylate, 3-ethoxycarbonyl piperidine, 3-piperidinecarboxylic acid, ethyl ester, piperidine-3-carboxylic acid ethyl ester, ethylnipecotate, a-ethyl nipecotate
CCOC(=O)C1CCCNC1
157.213
157.21
≥98.0% (GC,T)

Chemical Identifiers

5006-62-2
157.213
XIWBSOUNZWSFKU-UHFFFAOYSA-N
98969
CCOC(=O)C1CCCNC1
C8H15NO2
MFCD00005991
ethyl nipecotate, ethyl 3-piperidinecarboxylate, nipecotic acid ethyl ester, 3-piperidinecarboxylic acid ethyl ester, ethylpiperidine-3-carboxylate, 3-ethoxycarbonyl piperidine, 3-piperidinecarboxylic acid, ethyl ester, piperidine-3-carboxylic acid ethyl ester, ethylnipecotate, a-ethyl nipecotate
ethyl piperidine-3-carboxylate
Safety and Handling

Safety and Handling

TSCA : No

Documents