missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 3-Phenylpropionate 98.0+%, TCI America™
Supplier: TCI America P1304250G
Specifications
| Ethyl 3-Phenylpropionate | |
| Colorless | |
| C11H14O2 | |
| 250 g | |
| JAGZUIGGHGTFHO-UHFFFAOYSA-N | |
| ethyl 3-phenylpropanoate | |
| 16237 | |
| ≥98.0% (GC) |
| 2021-28-5 | |
| 248°C | |
| MFCD00009206 | |
| ethyl 3-phenylpropionate, ethyl hydrocinnamate, ethyl dihydrocinnamate, benzenepropanoic acid, ethyl ester, 3-phenylpropionic acid ethyl ester, hydrocinnamic acid, ethyl ester, ethylhydrocinnamoate, ethyl benzenepropanoate, unii-58e9z9v64d, ethylphenyl propanoate | |
| CCOC(=O)CCC1=CC=CC=C1 | |
| 178.231 | |
| 178.23 | |
| Liquid |
Chemical Identifiers
| 2021-28-5 | |
| 178.231 | |
| JAGZUIGGHGTFHO-UHFFFAOYSA-N | |
| 16237 | |
| CCOC(=O)CCC1=CC=CC=C1 |
| C11H14O2 | |
| MFCD00009206 | |
| ethyl 3-phenylpropionate, ethyl hydrocinnamate, ethyl dihydrocinnamate, benzenepropanoic acid, ethyl ester, 3-phenylpropionic acid ethyl ester, hydrocinnamic acid, ethyl ester, ethylhydrocinnamoate, ethyl benzenepropanoate, unii-58e9z9v64d, ethylphenyl propanoate | |
| ethyl 3-phenylpropanoate |
Safety and Handling
EINECSNumber : (3)-1730
TSCA : Yes