Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 3-oxo-3-(4-pyridyl)propionate, 98%

Chemical Identifiers

Chemical Name or Material Ethyl 3-oxo-3-(4-pyridyl)propionate
CAS 26377-17-3
Formula Weight 193.2
MDL Number MFCD00094021
Synonym Ethyl isonicotinoylacetate; 3-Oxo-3-(4-pyridyl)propionic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6164706

 
alfa aesar™
H6164706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6164714

 
alfa aesar™
H6164714
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 193.2
Melting Point 53° to 59°C
Chemical Name or Material Ethyl 3-oxo-3-(4-pyridyl)propionate
Refractive Index 1.5100
Physical Form Solid
Assay 98%
MDL Number MFCD00094021
CAS 26377-17-3
Synonym Ethyl isonicotinoylacetate; 3-Oxo-3-(4-pyridyl)propionic acid ethyl ester
Density 1.155g/mL
Color Pale brown
Quantity 5g