Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 3-oxo-3-(2-pyridyl)propionate, 96%

Chemical Identifiers

Chemical Name or Material Ethyl 3-oxo-3-(2-pyridyl)propionate
CAS 26510-52-1
Formula Weight 193.2
MDL Number MFCD00094022
Beilstein 145706
Synonym 3-Oxo-3-(2-pyridyl)propionic acid ethyl ester; Ethyl picolinoylacetate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6453303

  SDS
alfa aesar™
H6453303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6453306

  SDS
alfa aesar™
H6453306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH64533MD

  SDS
alfa aesar™
H64533MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Boiling Point 100° to 104°C (0.3mmHg)
Formula Weight 193.2
CAS 26510-52-1
Refractive Index 1.5160
Synonym 3-Oxo-3-(2-pyridyl)propionic acid ethyl ester; Ethyl picolinoylacetate
Assay 96%
Health Hazard 1 Warning
Beilstein 145706
Density 1.164g/mL
MDL Number MFCD00094022
Chemical Name or Material Ethyl 3-oxo-3-(2-pyridyl)propionate
Quantity 1g
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Physical Form Liquid
Sensitivity Light Sensitive
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.