Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 3-nitrocinnamate, 98%

$59.38 - $215.47

Chemical Identifiers

Chemical Name or Material Ethyl 3-nitrocinnamate
CAS 5396-71-4
Molecular Formula C11H11NO4
Formula Weight 221.21
MDL Number MFCD00017027
Beilstein 2618102
Synonym 3-Nitrocinnamic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1299506

  SDS
alfa aesar™
A1299506
5g Each for $59.38

AAA1299514

  SDS
alfa aesar™
A1299514
25g Each for $215.47
Description & Specifications

Specifications

Formula Weight 221.21
Quantity 5g
CAS 5396-71-4
Chemical Name or Material Ethyl 3-nitrocinnamate
Assay 98%
Synonym 3-Nitrocinnamic acid ethyl ester
Beilstein 2618102
Melting Point 74° to 76°C
Molecular Formula C11H11NO4
MDL Number MFCD00017027