Please login to your online account to display your discounted pricing

Ethyl 3-(N,N-dimethylamino)acrylate, 99+%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Ethyl 3-(N, N-dimethylamino)acrylate
Name Note 99+%
CAS 924-99-2
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula C7H13NO2
Linear Formula (CH3)2NCH=CHCOOC2H5
Formula Weight 143.19
MDL Number MFCD00144269
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC269630250

  SDS
fisher scientific acros
269630250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥99.0%
Formula Weight 143.19
Physical Form Liquid
Quantity 25g
Color Clear yellow to light orange
Density 1g/mL
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
May cause an allergic skin reaction.
Health Hazard 3 GHS P Statement:
Wear eye protection/face protection.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Refractive Index 1.5105 to 1.5125 (20°C, 589nm)
Packaging Glass bottle
Assay Percent Range 99+%
Chemical Name or Material Ethyl 3-(N, N-dimethylamino)acrylate
MDL Number MFCD00144269
CAS 924-99-2
Name Note 99+%
Molecular Formula C7H13NO2
Linear Formula (CH3)2NCH=CHCOOC2H5
Safety and Handling

GHS P Statement:
Wear eye protection/face protection.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause an allergic skin reaction.