Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 3-methylvalerate, 98%

Chemical Identifiers

Chemical Name or Material Ethyl 3-methylvalerate
CAS 5870-68-8
Molecular Formula C8H16O2
Formula Weight 144.21
MDL Number MFCD00041390
UN Number UN3272
Synonym Ethyl 3-methylpentanoate; 3-Methylvaleric acid ethyl esterer
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1886806

 
alfa aesar™
A1886806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1886814

 
alfa aesar™
A1886814
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 144.21
MDL Number MFCD00041390
UN Number UN3272
Synonym Ethyl 3-methylpentanoate; 3-Methylvaleric acid ethyl esterer
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H226
Flammable liquid and vapour.
Health Hazard 3 GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.
Flash Point 45°C (113°F)
Odor Ester-like
Quantity 5g
Chemical Name or Material Ethyl 3-methylvalerate
Assay 98%
Molecular Formula C8H16O2
CAS 5870-68-8
Boiling Point 158°C
Safety and Handling

Warning

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226
Flammable liquid and vapour.