Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 3-(methylthio)propionate, 98%

$51.38 - $632.84

Chemical Identifiers

Chemical Name or Material Ethyl 3- (methylthio)propionate
CAS 13327-56-5
Molecular Formula CH3SCH2CH2CO2CH2CH3
Formula Weight 148.23
MDL Number MFCD00039900
Beilstein 1748688
Synonym 3-(Methylthio)propionic acid ethyl ester; Ethyl 3-(methylmercapto)propionate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1174214

  SDS
alfa aesar™
A1174214
25g Each for $51.38

AAA1174222

  SDS
alfa aesar™
A1174222
100g Each for $149.18

AAA1174236

  SDS
alfa aesar™
A1174236
500g Each for $632.84
Description & Specifications

Specifications

Formula Weight 148.23
Chemical Name or Material Ethyl 3- (methylthio)propionate
Assay 98%
Beilstein 1748688
Health Hazard 1 Warning
CAS 13327-56-5
Synonym 3-(Methylthio)propionic acid ethyl ester; Ethyl 3-(methylmercapto)propionate
TSCA TSCA
Boiling Point 197°C
Health Hazard 2 GHS H Statement
H227
Combustible liquid.
Health Hazard 3 GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 25g
Molecular Formula CH3SCH2CH2CO2CH2CH3
MDL Number MFCD00039900
Flash Point 79°C (174°F)
Safety and Handling

Warning

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H227
Combustible liquid.