missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 3-(methylthio)propionate, 98%

Manufacturer:  Alfa Aesar™ A1174222

 View more versions of this product

Catalog No. AAA1174222



Specifications

Specifications

Ethyl 3- (methylthio)propionate
98%
MFCD00039900
3-(Methylthio)propionic acid ethyl ester; Ethyl 3-(methylmercapto)propionate
CCOC(=O)CCSC
148.22
CHEBI:87503
197°C
79°C (174°F)
13327-56-5
C6H12O2S
1748688
YSNWHRKJEKWJNY-UHFFFAOYSA-N
ethyl 3-methylsulfanylpropanoate
61592
148.23
100g

Chemical Identifiers

13327-56-5
148.22
YSNWHRKJEKWJNY-UHFFFAOYSA-N
61592
ethyl 3-methylsulfanylpropanoate
C6H12O2S
MFCD00039900
3-(Methylthio)propionic acid ethyl ester; Ethyl 3-(methylmercapto)propionate
CHEBI:87503
CCOC(=O)CCSC
Safety and Handling

Safety and Handling

GHS H Statement
H227
Combustible liquid.

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

SDS Documents