missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl 3-Methylcrotonate, TCI America™

$45.83 - $132.92

Chemical Identifiers

CAS 638-10-8
Molecular Formula C7H12O2
Molecular Weight (g/mol) 128.171
MDL Number MFCD00009187
InChI Key UTXVCHVLDOLVPC-UHFFFAOYSA-N
Synonym 3,3-Dimethylacrylic Acid Ethyl Ester, Ethyl 3,3-Dimethylacrylate, Ethyl 3-Methyl-2-butenoate, 3-Methylcrotonic Acid Ethyl Ester, 3-Methyl-2-butenoic Acid Ethyl Ester
PubChem CID 12516
IUPAC Name ethyl 3-methylbut-2-enoate
SMILES CCOC(=O)C=C(C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M144125ML View Documents TCI America
M144125ML
25mL Each for $132.92
 
M14415ML View Documents TCI America
M14415ML
5mL Each for $45.83
 
Specifications

Chemical Identifiers

638-10-8
128.171
UTXVCHVLDOLVPC-UHFFFAOYSA-N
12516
CCOC(=O)C=C(C)C
C7H12O2
MFCD00009187
3,3-Dimethylacrylic Acid Ethyl Ester, Ethyl 3,3-Dimethylacrylate, Ethyl 3-Methyl-2-butenoate, 3-Methylcrotonic Acid Ethyl Ester, 3-Methyl-2-butenoic Acid Ethyl Ester
ethyl 3-methylbut-2-enoate

Specifications

Colorless
638-10-8
MFCD00009187
3,3-Dimethylacrylic Acid Ethyl Ester, Ethyl 3,3-Dimethylacrylate, Ethyl 3-Methyl-2-butenoate, 3-Methylcrotonic Acid Ethyl Ester, 3-Methyl-2-butenoic Acid Ethyl Ester
CCOC(=O)C=C(C)C
128.171
128.17
≥97.0% (GC)
25mL
C7H12O2
3272
UTXVCHVLDOLVPC-UHFFFAOYSA-N
ethyl 3-methylbut-2-enoate
12516
Clear Liquid at 20°C
Ethyl 3-Methylcrotonate
Safety and Handling

Safety and Handling

RTECSNumber : GQ5750000

TSCA : Yes