Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 3-methyl-4H-1,4-benzothiazine-2-carboxylate, 98%

$38.18 - $145.31

Chemical Identifiers

Chemical Name or Material Ethyl 3-methyl-4H-1,4-benzothiazine-2-carboxylate
CAS 7625-01-6
Molecular Formula C12H13NO2S
Formula Weight 235.31
MDL Number MFCD00173950
Synonym 3-Methyl-4H-1,4-benzothiazine-2-carboxylic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1836003

 
alfa aesar™
A1836003
1g Each for $38.18

AAA1836006

 
alfa aesar™
A1836006
5g Each for $145.31
Description & Specifications

Specifications

Formula Weight 235.31
Chemical Name or Material Ethyl 3-methyl-4H-1,4-benzothiazine-2-carboxylate
Assay 98%
Molecular Formula C12H13NO2S
MDL Number MFCD00173950
CAS 7625-01-6
Synonym 3-Methyl-4H-1,4-benzothiazine-2-carboxylic acid ethyl ester
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 142° to 144°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.