Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 3-methyl-4-nitrobenzoate, 98%

$43.84 - $662.97

Chemical Identifiers

Chemical Name or Material Ethyl 3-methyl-4-nitrobenzoate
CAS 30650-90-9
Molecular Formula C10H11NO4
Formula Weight 209.2
MDL Number MFCD00115804
Synonym 3-Methyl-4-nitrobenzoic acid ethyl ester; Ethyl 4-nitro-m-toluate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1862509

  SDS
alfa aesar™
A1862509
10g Each for $43.84

AAA1862530

  SDS
alfa aesar™
A1862530
250g Each for $662.97

AAA1862518

  SDS
alfa aesar™
A1862518
50g Each for $159.72
Description & Specifications

Specifications

Formula Weight 209.2
CAS 30650-90-9
Synonym 3-Methyl-4-nitrobenzoic acid ethyl ester; Ethyl 4-nitro-m-toluate
Melting Point 53° to 55°C
Chemical Name or Material Ethyl 3-methyl-4-nitrobenzoate
Assay 98%
MDL Number MFCD00115804
Quantity 10g
Molecular Formula C10H11NO4