missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl 3-Methyl-3-phenylglycidate (mixture of isomers), TCI America™

Manufacturer:  TCI America E0873250G

 View more versions of this product

Catalog No. E0873250G



Specifications

Specifications

Ethyl 3-Methyl-3-phenylglycidate (mixture of isomers)
Yellow
77-83-8
MFCD00022338
LQKRYVGRPXFFAV-UHFFFAOYSA-N
ethyl 3-methyl-3-phenyloxirane-2-carboxylate
6501
Clear Liquid at 20°C
86°C
250g
C12H14O3
ethyl methylphenylglycidate, ethyl 3-methyl-3-phenylglycidate, strawberry aldehyde, fraeseol, empg, c-16 aldehyde, aldehyde c-16, ethyl methylphenyl glycidate, ethyl-3-methyl-3-phenylglycidate, ethyl 2,3-epoxy-3-phenylbutyrate
CCOC(=O)C1C(O1)(C)C2=CC=CC=C2
206.241
206.24
≥95.0% (GC)

Chemical Identifiers

77-83-8
206.241
LQKRYVGRPXFFAV-UHFFFAOYSA-N
6501
CCOC(=O)C1C(O1)(C)C2=CC=CC=C2
C12H14O3
MFCD00022338
ethyl methylphenylglycidate, ethyl 3-methyl-3-phenylglycidate, strawberry aldehyde, fraeseol, empg, c-16 aldehyde, aldehyde c-16, ethyl methylphenyl glycidate, ethyl-3-methyl-3-phenylglycidate, ethyl 2,3-epoxy-3-phenylbutyrate
ethyl 3-methyl-3-phenyloxirane-2-carboxylate
Safety and Handling

Safety and Handling

EINECSNumber : (3)-3096

RTECSNumber : MW5250000

TSCA : Yes

Documents