Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 3-methyl-3-phenylglycidate, cis + trans, 98%

$93.27 - $292.31

Chemical Identifiers

Chemical Name or Material Ethyl 3-methyl-3-phenylglycidate
CAS 77-83-8
Name Note cis + trans
Molecular Formula C12H14O3
Formula Weight 206.24
MDL Number MFCD00022338
Beilstein 12299
Synonym 3-Methyl-3-phenylglycidic acid ethyl ester; 3-Phenyl-2,3-epoxybutyric acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AAB236540B

  SDS
alfa aesar™
B236540B
Each for $292.31

AAB2365430

  SDS
alfa aesar™
B2365430
Each for $93.27
Description & Specifications

Specifications

Formula Weight 206.24
Chemical Name or Material Ethyl 3-methyl-3-phenylglycidate
TSCA TSCA
Name Note cis + trans
Assay 98%
Molecular Formula C12H14O3
MDL Number MFCD00022338
CAS 77-83-8
Beilstein 12299
Synonym 3-Methyl-3-phenylglycidic acid ethyl ester; 3-Phenyl-2,3-epoxybutyric acid ethyl ester
Boiling Point 272° to 275°C
Quantity 1000g
Flash Point 134°C (273°F)
RTECS Number MW5250000