Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 3-methyl-2-thionoimidazoline-1-carboxylate, 97%


Quantity 5g

Manufacturer: alfa aesar™  L0821806

 View more versions of this product

Catalog No. AAL0821806

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 186.24
Chemical Name or Material Ethyl 3-methyl-2-thionoimidazoline-1-carboxylate
Assay 97%
Molecular Formula C7H10N2O2S
MDL Number MFCD00027421
CAS 22232-54-8
Beilstein 144339
Synonym Carbimazole; 3-Methyl-2-thionoimidazoline-1-carboxylic acid ethyl ester
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H319-H317
Harmful if swallowed.
Causes serious eye irritation.
May cause an allergic skin reaction.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P301+P312-P363-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wash contaminated clothing before reuse.
Dispose of contents/container in accordance with local/regional/national/international regulations.
pH 5.0 to 7.0
Melting Point 122° to 126°C
Quantity 5g
RTECS Number NJ2441000
Merck Index 14,1798

Specifications

Chemical Name or Material Ethyl 3-methyl-2-thionoimidazoline-1-carboxylate
CAS 22232-54-8
Molecular Formula C7H10N2O2S
Formula Weight 186.24
MDL Number MFCD00027421
Beilstein 144339
Merck Index 14,1798
Synonym Carbimazole; 3-Methyl-2-thionoimidazoline-1-carboxylic acid ethyl ester

Applications
Ethyl 3-methyl-2-thionoimidazoline-1-carboxylate is used as thyroid inhibitor.

Solubility
Soluble in water, ethanol, chloroform and acetone. Sightly soluble in water.

Notes
Light sensitive. Keep the container tightly closed in a dry and well-ventilated place.

Accessories

Warning

GHS P Statement
P261-P280-P305+P351+P338-P301+P312-P363-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wash contaminated clothing before reuse.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H319-H317
Harmful if swallowed.
Causes serious eye irritation.
May cause an allergic skin reaction.