Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 3-methyl-1H-pyrazole-5-carboxylate, 97%

$59.22 - $760.92

Chemical Identifiers

Chemical Name or Material Ethyl 3-methyl-1H-pyrazole-5-carboxylate
CAS 4027-57-0
Molecular Formula C7H10N2O2
Formula Weight 154.17
MDL Number MFCD00052514
Beilstein 4192
Synonym 3-Methyl-1H-pyrazole-5-carboxylic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1230403

  SDS
alfa aesar™
A1230403
1g Each for $59.22

AAA1230406

  SDS
alfa aesar™
A1230406
5g Each for $197.39

AAA1230414

  SDS
alfa aesar™
A1230414
25g Each for $760.92
Description & Specifications

Specifications

Formula Weight 154.17
Chemical Name or Material Ethyl 3-methyl-1H-pyrazole-5-carboxylate
Assay 97%
Molecular Formula C7H10N2O2
MDL Number MFCD00052514
CAS 4027-57-0
Beilstein 4192
Synonym 3-Methyl-1H-pyrazole-5-carboxylic acid ethyl ester
Melting Point 80° to 83°C
Quantity 1g