missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl 3-Methoxypropionate 99.0+%, TCI America™

Supplier:  TCI America M100125G

 View more versions of this product

Catalog No. M100125G



Description

Description

Specifications

Specifications

Ethyl 3-Methoxypropionate
Colorless
C6H12O3
25 g
3-Methoxypropionic Acid Ethyl Ester
CCOC(=O)CCOC
132.16
132.16
Liquid
10606-42-5
158°C
MFCD00143172
3272
IJUHLFUALMUWOM-UHFFFAOYSA-N
ethyl 3-methoxypropanoate
82767
≥99.0% (GC)

Chemical Identifiers

10606-42-5
132.16
IJUHLFUALMUWOM-UHFFFAOYSA-N
82767
CCOC(=O)CCOC
C6H12O3
MFCD00143172
3-Methoxypropionic Acid Ethyl Ester
ethyl 3-methoxypropanoate
Safety and Handling

Safety and Handling

EINECSNumber : (2)-1350&(2)-1379

RTECSNumber : UF5273000

TSCA : No

SDS
Documents

Documents

Promotions

Promotions