missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ethyl 3-methoxyphenylacetate, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1507014
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Ethyl 3-methoxyphenylacetate | |
109°C to 110°C (1 mmHg) | |
MFCD00026897 | |
ethyl 3-methoxyphenylacetate, ethyl 2-3-methoxyphenyl acetate, ethyl 3-methoxyphenyl acetate, benzeneacetic acid, 3-methoxy-, ethyl ester, acmc-20akzh, 3-methoxy-phenyl-acetic acid ethyl ester, ethyl 2-3-methoxyphenyl ethanoate, 3-methoxyphenylacetic acid ethyl ester, 3-methoxybenzeneacetic acid ethyl ester, 2-3-methoxyphenyl acetic acid ethyl ester | |
CCOC(=O)CC1=CC(=CC=C1)OC | |
194.23 | |
194.23 |
35553-92-5 | |
C11H14O3 | |
25 g | |
XXVVNHCWPHMLEZ-UHFFFAOYSA-N | |
ethyl 2-(3-methoxyphenyl)acetate | |
2774971 | |
98% |
Chemical Identifiers
35553-92-5 | |
194.23 | |
XXVVNHCWPHMLEZ-UHFFFAOYSA-N | |
2774971 | |
CCOC(=O)CC1=CC(=CC=C1)OC |
C11H14O3 | |
MFCD00026897 | |
ethyl 3-methoxyphenylacetate, ethyl 2-3-methoxyphenyl acetate, ethyl 3-methoxyphenyl acetate, benzeneacetic acid, 3-methoxy-, ethyl ester, acmc-20akzh, 3-methoxy-phenyl-acetic acid ethyl ester, ethyl 2-3-methoxyphenyl ethanoate, 3-methoxyphenylacetic acid ethyl ester, 3-methoxybenzeneacetic acid ethyl ester, 2-3-methoxyphenyl acetic acid ethyl ester | |
ethyl 2-(3-methoxyphenyl)acetate |
Safety and Handling
EINECSNumber : 252-614-5
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only