Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 3-methoxybenzoate, 98+%

$58.81 - $929.73

Chemical Identifiers

Chemical Name or Material Ethyl 3-methoxybenzoate
CAS 10259-22-0
Molecular Formula C10H12O3
Formula Weight 180.2
MDL Number MFCD00015149
Beilstein 2576420
Synonym Ethyl m-anisate; 3-Methoxybenzoic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1090209

 
alfa aesar™
A1090209
10g Each for $58.81

AAA1090230

 
alfa aesar™
A1090230
250g Each for $929.73

AAA1090218

 
alfa aesar™
A1090218
50g Each for $226.27
Description & Specifications

Specifications

Formula Weight 180.2
Chemical Name or Material Ethyl 3-methoxybenzoate
Assay ≥98%
Molecular Formula C10H12O3
MDL Number MFCD00015149
CAS 10259-22-0
Beilstein 2576420
Synonym Ethyl m-anisate; 3-Methoxybenzoic acid ethyl ester
Boiling Point 250° to 252°C
Quantity 10g