missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl 3-Mercaptopropionate 98.0+%, TCI America™

Manufacturer:  TCI America M1038100G

 View more versions of this product

Catalog No. M1038100G



Specifications

Specifications

Ethyl 3-Mercaptopropionate
100g
C5H10O2S
3-mercaptopropanoic acid ethyl ester
CCOC(=O)CCS
134.19
134.19
≥98.0% (GC)
Colorless
5466-06-8
MFCD00004896
CJQWLNNCQIHKHP-UHFFFAOYSA-N
ethyl 3-sulfanylpropanoate
21625
Clear Liquid at 20°C

Chemical Identifiers

5466-06-8
134.19
CJQWLNNCQIHKHP-UHFFFAOYSA-N
21625
CCOC(=O)CCS
C5H10O2S
MFCD00004896
3-mercaptopropanoic acid ethyl ester
ethyl 3-sulfanylpropanoate
Safety and Handling

Safety and Handling

TSCA : Yes

Documents