Learn More
Ethyl 3-isothiocyanatopropionate, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L1091103
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Ethyl 3-isothiocyanatopropionate | |
1.12 | |
Pungent | |
1.5014 | |
MFCD00041130 | |
UN2922 | |
Moisture sensitive | |
UPTRONYNXNYITM-UHFFFAOYSA-N | |
ethyl 3-isothiocyanatopropanoate | |
526514 | |
97% |
17126-62-4 | |
75°C to 76°C (8 mmHg) | |
C6H9NO2S | |
SCNCH2CH2CO2CH2CH3 | |
1 g | |
1705633 | |
ethyl 3-isothiocyanatopropionate, 3-isothiocyanato-propionic acid ethyl ester, acmc-1ca2j, ethyl-3-isothiocynato propionate, isothiocyanato-propionic acid ethyl ester, 3-isothiocyanatopropionic acid ethyl ester, propanoic acid,3-isothiocyanato-,ethyl ester, propanoic acid,3-isothiocyanato-, ethyl ester | |
CCOC(=O)CCN=C=S | |
159.203 | |
159.2 |
Chemical Identifiers
17126-62-4 | |
159.203 | |
UPTRONYNXNYITM-UHFFFAOYSA-N | |
526514 | |
CCOC(=O)CCN=C=S |
C6H9NO2S | |
MFCD00041130 | |
ethyl 3-isothiocyanatopropionate, 3-isothiocyanato-propionic acid ethyl ester, acmc-1ca2j, ethyl-3-isothiocynato propionate, isothiocyanato-propionic acid ethyl ester, 3-isothiocyanatopropionic acid ethyl ester, propanoic acid,3-isothiocyanato-,ethyl ester, propanoic acid,3-isothiocyanato-, ethyl ester | |
ethyl 3-isothiocyanatopropanoate |
Safety and Handling
GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302+H312-H314-H331-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only