missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl 3-Iodobenzoate 98.0+%, TCI America™

$46.08 - $134.97

Chemical Identifiers

CAS 58313-23-8
Molecular Formula C9H9IO2
Molecular Weight (g/mol) 276.073
MDL Number MFCD00044769
InChI Key POGCXCWRMMXDAQ-UHFFFAOYSA-N
Synonym 3-iodobenzoic acid ethyl ester
PubChem CID 143542
IUPAC Name ethyl 3-iodobenzoate
SMILES CCOC(=O)C1=CC(=CC=C1)I
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
I05365G View Documents Tci America
I05365G
5g Each for $46.08
 
I053625G View Documents Tci America
I053625G
25g Each for $134.97
 
Specifications

Chemical Identifiers

58313-23-8
276.073
POGCXCWRMMXDAQ-UHFFFAOYSA-N
143542
CCOC(=O)C1=CC(=CC=C1)I
C9H9IO2
MFCD00044769
3-iodobenzoic acid ethyl ester
ethyl 3-iodobenzoate

Specifications

161°C
5g
C9H9IO2
3-iodobenzoic acid ethyl ester
CCOC(=O)C1=CC(=CC=C1)I
276.073
276.07
≥98.0% (GC)
Yellow
58313-23-8
MFCD00044769
POGCXCWRMMXDAQ-UHFFFAOYSA-N
ethyl 3-iodobenzoate
143542
Clear Liquid at 20°C
Ethyl 3-Iodobenzoate
Safety and Handling

Safety and Handling

TSCA : No