Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 3-iodobenzoate, 99%

$79.12 - $1,259.14

Chemical Identifiers

Chemical Name or Material Ethyl 3-iodobenzoate
CAS 58313-23-8
Molecular Formula C9H9IO2
Formula Weight 276.08
MDL Number MFCD00044769
Beilstein 2089064
Synonym 3-Iodobenzoic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1421009

 
alfa aesar™
A1421009
10g Each for $79.12

AAA1421030

 
alfa aesar™
A1421030
250g Each for $1,259.14
Description & Specifications

Specifications

Formula Weight 276.08
Chemical Name or Material Ethyl 3-iodobenzoate
TSCA TSCA
Assay 99%
Molecular Formula C9H9IO2
MDL Number MFCD00044769
CAS 58313-23-8
Beilstein 2089064
Synonym 3-Iodobenzoic acid ethyl ester
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Flash Point >110°C (230°F)
Boiling Point 272°C
Quantity 10g

Applications
Ethyl 3-iodobenzoate is used as pharmaceutical intermediates.

Solubility
Not miscible in water.

Notes
Light Sensitive. Store in the dark. Store away from oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.