Please login to your online account to display your discounted pricing

Ethyl 3-iodobenzoate, 98%, ACROS Organics™


Quantity 50mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  250040500

 View more versions of this product

Catalog No. AC250040500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 276.07
Quantity 50mL
Density 1.6600g/mL
Chemical Name or Material Ethyl 3-iodobenzoate
Packaging Glass bottle
Name Note 98%
Assay Percent Range 98%
Molecular Formula C9H9IO2
Linear Formula IC6H4CO2CH2CH3
MDL Number MFCD00044769
CAS 58313-23-8
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Ethyl 3-iodobenzoate
Name Note 98%
CAS 58313-23-8
Assay Percent Range 98%
Molecular Formula C9H9IO2
Linear Formula IC6H4CO2CH2CH3
Formula Weight 276.07
MDL Number MFCD00044769
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

SDS & Safety Documents