Please login to your online account to display your discounted pricing

Ethyl 3-iodobenzoate, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Ethyl 3-iodobenzoate
Name Note 98%
CAS 58313-23-8
Assay Percent Range 98%
Molecular Formula C9H9IO2
Linear Formula IC6H4CO2CH2CH3
Formula Weight 276.07
MDL Number MFCD00044769
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC250040500

  SDS
thermo fisher scientific
250040500
Glass bottle 50mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 276.07
Quantity 50mL
Density 1.6600g/mL
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Name Note 98%
MDL Number MFCD00044769
Assay Percent Range 98%
Chemical Name or Material Ethyl 3-iodobenzoate
Molecular Formula C9H9IO2
Linear Formula IC6H4CO2CH2CH3
CAS 58313-23-8
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.