missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl 3-hydroxybutyrate, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 118541000

 View more versions of this product

Catalog No. AC118541000



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Ethyl 3-hydroxybutyrate
5405-41-4
1.0100g/mL
64°C
98.5% min. (GC)
C6H12O3
CH3CH(OH)CH2CO2C2H5
100 g
14,162
ethyl 3-hydroxybutyrate, ethyl beta-hydroxybutyrate, butanoic acid, 3-hydroxy-, ethyl ester, butyric acid, 3-hydroxy-, ethyl ester, fema no. 3428, ethyl 3-hydroxybutyrate natural, ethyl dl-3-hydroxybutyrate, ethyl 1-3-hydroxybutyrate, dl-3-hydroxy-n-butyric acid ethyl ester, ethyl3-hydroxybutyrate
OMSUIQOIVADKIM-UHFFFAOYNA-N
ethyl 3-hydroxybutanoate
62572
132.16
Liquid
99%
Colorless
170°C
Authentic
Glass bottle
1.4195 to 1.4215
MFCD00004545
03, 309
1.01
Solubility in water: soluble
CCOC(=O)CC(C)O
132.16
CHEBI:87685
99%

Chemical Identifiers

5405-41-4
132.16
OMSUIQOIVADKIM-UHFFFAOYNA-N
CHEBI:87685
CCOC(=O)CC(C)O
C6H12O3
MFCD00004545
62572
ethyl 3-hydroxybutanoate
Safety and Handling

Safety and Handling

EINECSNumber : 226-456-2

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only