Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 3-hydroxybenzoate, 99%

$69.76 - $714.20

Chemical Identifiers

Chemical Name or Material Ethyl 3-hydroxybenzoate
CAS 7781-98-8
Molecular Formula C9H10O3
Formula Weight 166.18
MDL Number MFCD00002296
Beilstein 2208566
Synonym 3-Hydroxybenzoic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1534314

  SDS
alfa aesar™
A1534314
25g Each for $69.76

AAA1534322

  SDS
alfa aesar™
A1534322
100g Each for $183.82

AAA1534336

  SDS
alfa aesar™
A1534336
500g Each for $714.20
Description & Specifications

Specifications

Formula Weight 166.18
Chemical Name or Material Ethyl 3-hydroxybenzoate
Assay 99%
Molecular Formula C9H10O3
MDL Number MFCD00002296
CAS 7781-98-8
Beilstein 2208566
Synonym 3-Hydroxybenzoic acid ethyl ester
Boiling Point 187° to 188°C (31mmHg)
Health Hazard 1 Warning
Melting Point 72° to 76°C
Quantity 25g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Applications
Ethyl 3-hydroxybenzoate has been used in the synthesis of chiral derivatives of ethyl 3-(2-methylbutyloxy)benzoate and 4-(2-methylbutyloxy)-4′-acetylbiphenyl, via Mitsunobu reaction. It is a food-grade antimicrobial agent.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.