Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 3-hydroxy-3-phenylpropionate, 95%

Chemical Identifiers

Chemical Name or Material Ethyl 3-hydroxy-3-phenylpropionate
CAS 5764-85-2
Formula Weight 194.23
MDL Number MFCD00086891
Beilstein 2211602
Synonym 3-Hydroxy-3-phenylpropionic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6570803

 
alfa aesar™
H6570803
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH65708MD

 
alfa aesar™
H65708MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6570806

 
alfa aesar™
H6570806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 194.23
Chemical Name or Material Ethyl 3-hydroxy-3-phenylpropionate
Density 1.088g/mL
TSCA TSCA
Physical Form Solid
Refractive Index 1.5080
MDL Number MFCD00086891
CAS 5764-85-2
Synonym 3-Hydroxy-3-phenylpropionic acid ethyl ester
Assay 95%
Beilstein 2211602
Quantity 1g