missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ethyl 3-hydroxy-3-phenylpropionate, 95%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals H65708MD
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Ethyl 3-hydroxy-3-phenylpropionate | |
1.088 g/mL | |
1.508 | |
250 mg | |
ethyl 3-hydroxy-3-phenylpropionate, 3-hydroxy-3-phenylpropionic acid ethyl ester, hydracrylic acid, 3-phenyl-, ethyl ester, 3-hydroxy-3-phenyl-propionic acid ethyl ester, benzenepropanoic acid, .beta.-hydroxy-, ethyl ester, 3-hydroxy-3-phenyl-propionicacidethylester, ethyl-3-hydroxy-3-phenyl propionate, ethyl 3-hydroxy-3-phenyl-propanoate, ethyl 3-phenylhydracrylate, +/-, acmc-20apru | |
CCOC(=O)CC(C1=CC=CC=C1)O | |
194.23 | |
194.23 | |
Solid |
5764-85-2 | |
C11H14O3 | |
MFCD00086891 | |
2211602 | |
DVIBDQWVFHDBOP-UHFFFAOYSA-N | |
ethyl 3-hydroxy-3-phenylpropanoate | |
99818 | |
95% |
Chemical Identifiers
5764-85-2 | |
194.23 | |
DVIBDQWVFHDBOP-UHFFFAOYSA-N | |
99818 | |
CCOC(=O)CC(C1=CC=CC=C1)O |
C11H14O3 | |
MFCD00086891 | |
ethyl 3-hydroxy-3-phenylpropionate, 3-hydroxy-3-phenylpropionic acid ethyl ester, hydracrylic acid, 3-phenyl-, ethyl ester, 3-hydroxy-3-phenyl-propionic acid ethyl ester, benzenepropanoic acid, .beta.-hydroxy-, ethyl ester, 3-hydroxy-3-phenyl-propionicacidethylester, ethyl-3-hydroxy-3-phenyl propionate, ethyl 3-hydroxy-3-phenyl-propanoate, ethyl 3-phenylhydracrylate, +/-, acmc-20apru | |
ethyl 3-hydroxy-3-phenylpropanoate |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 227-288-2
TSCA : Yes
Recommended Storage : Ambient temperatures