Please login to your online account to display your discounted pricing

Ethyl 3-furoate, 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Ethyl 3-furoate
Name Note 99%
CAS 614-98-2
Assay Percent Range 99%
Molecular Formula C7H8O3
Formula Weight 140.14
MDL Number MFCD00005348
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC269030050

  SDS
fisher scientific acros
269030050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC269030010

  SDS
thermo fisher scientific
269030010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 140.14
Quantity 5g
EINECS Number 210-403-5
Packaging Glass bottle
Name Note 99%
Molecular Formula C7H8O3
CAS 614-98-2
Chemical Name or Material Ethyl 3-furoate
Assay Percent Range 99%
MDL Number MFCD00005348