missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl 3-furoate, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 269030010

 View more versions of this product

Catalog No. AC269030010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Ethyl3-furoate
59°C
1.4600 to 1.462
1g
614-98-2
C7H8O3
18,4052
LOFDXZJSDVCYAS-UHFFFAOYSA-N
ethyl furan-3-carboxylate
69201
99%
1.1100g/mL
Glass bottle
1.11
99%
98.5% min. (GC)
MFCD00005348
3-furancarboxylic acid ethyl ester, 3-furancarboxylic acid, ethyl ester, acmc-1bczk, ethyl 3-furancarboxylate, ethyl 3-furoate, ethyl 3furoate, ethyl3-furancarboxylate, ethyl3-furoate, pubchem19999, unii-i8q0yn919k
CCOC(=O)C1=COC=C1
140.14
140.14

Chemical Identifiers

614-98-2
140.14
LOFDXZJSDVCYAS-UHFFFAOYSA-N
69201
CCOC(=O)C1=COC=C1
C7H8O3
MFCD00005348
3-furancarboxylic acid ethyl ester, 3-furancarboxylic acid, ethyl ester, acmc-1bczk, ethyl 3-furancarboxylate, ethyl 3-furoate, ethyl 3furoate, ethyl3-furancarboxylate, ethyl3-furoate, pubchem19999, unii-i8q0yn919k
ethyl furan-3-carboxylate
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

EINECSNumber : 210-403-5

SDS Documents