Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 3-(furfurylthio)propionate, 97%

$63.57 - $180.09

Chemical Identifiers

Chemical Name or Material Ethyl 3-(furfurylthio)propionate
CAS 94278-27-0
Molecular Formula C10H14O3S
Formula Weight 214.29
MDL Number MFCD00051819
Beilstein 4311671
Synonym Ethyl 3-(furfurylmercapto)propionate; 3-(Furfurylthio)propionic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2176609

  SDS
alfa aesar™
B2176609
10g Each for $63.57

AAB2176618

  SDS
alfa aesar™
B2176618
50g Each for $180.09
Description & Specifications

Specifications

Formula Weight 214.29
Chemical Name or Material Ethyl 3-(furfurylthio)propionate
Assay 97%
Molecular Formula C10H14O3S
MDL Number MFCD00051819
CAS 94278-27-0
Beilstein 4311671
Synonym Ethyl 3-(furfurylmercapto)propionate; 3-(Furfurylthio)propionic acid ethyl ester
Boiling Point 120° to 122°C (0.5mmHg)
Quantity 10g
Flash Point 99°C (210°F)