Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl (3-formylphenoxy)acetate, 96%

$105.03 - $357.11

Chemical Identifiers

Chemical Name or Material Ethyl (3-formylphenoxy)acetate
CAS 51264-68-7
Molecular Formula C11H12O4
Formula Weight 208.22
MDL Number MFCD04227168
Beilstein 2109732
Synonym (3-Formylphenoxy)acetic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL2039906

  SDS
alfa aesar™
L2039906
5g Each for $105.03

AAL2039914

  SDS
alfa aesar™
L2039914
25g Each for $357.11
Description & Specifications

Specifications

Formula Weight 208.22
CAS 51264-68-7
Health Hazard 1 Warning
Molecular Formula C11H12O4
Chemical Name or Material Ethyl (3-formylphenoxy)acetate
MDL Number MFCD04227168
Assay 96%
Quantity 5g
Synonym (3-Formylphenoxy)acetic acid ethyl ester
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Boiling Point 148° to 150°C (2mmHg)
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Beilstein 2109732
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.