Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 3-formylbenzoate, 98%

$128.22 - $441.77

Chemical Identifiers

Chemical Name or Material Ethyl 3-formylbenzoate
CAS 33745-47-0
Molecular Formula C10H10O3
Formula Weight 178.19
MDL Number MFCD08064275
Synonym 3-Ethoxycarbonylbenzaldehyde; 3-Formylbenzoic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5094503

  SDS
alfa aesar™
H5094503
1g Each for $441.77

AAH50945MD

  SDS
alfa aesar™
H50945MD
250mg Each for $128.22
Description & Specifications

Specifications

Formula Weight 178.19
Molecular Formula C10H10O3
MDL Number MFCD08064275
Synonym 3-Ethoxycarbonylbenzaldehyde; 3-Formylbenzoic acid ethyl ester
Quantity 1g
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material Ethyl 3-formylbenzoate
Assay 98%
CAS 33745-47-0
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

Applications
Ethyl 3-formylbenzoate is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.